Salisbury PRO-WEAR™ Arc Flash Personal Protection Equipment Kits
ATPV Rating: 40 cal/cm2
Manufacturer: WH Salisbury & Co. SK40L
 View more versions of this product
Catalog No. 19-138-167C
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120
    Material Indura™ Ultra soft™ Fabric
    Includes Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag
    Size Large
    Arc Flash Rating 40 Calories/cm2
  • Description & Specifications
Related Products
Product Content Feedback