Learn More
Salisbury PRO-WEAR™ Arc Flash Personal Protection Equipment Kits
ATPV Rating: 40 cal/cm2
Manufacturer:  WH Salisbury & Co. SK40XL
 View more versions of this product
Catalog No. 19-138-167D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F1506, NFPA 70E™, CE 0120
Indura™ Ultra soft™ Fabric
X-Large
40 Calories/cm2
Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag