Learn More
Salisbury PRO-WEAR™ Arc Flash Protection Coverall Kits
ATPV rating: 10 cal/cm2
Manufacturer:  WH Salisbury & Co. SKCA112XL
 View more versions of this product
Catalog No. 19-139-785
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NFPA 70E™ 2009, CE 0120
12 Calories/cm2
AS1000 face shield, hard hat, Salisbury storage bag and safety Glasses
2X-Large