Learn More
Honeywell Salisbury™ PRO-WEAR™ Flash Protection Bib Overalls
11 cal/cm2 to 100 cal/cm2 ATPV ratings
$567.41 - $1993.00
Specifications
Certifications/Compliance ASTM F1506, NFPA 70E™
Closure Type Adjustable Hook and Pile Waist Straps
For Use With (Application) Flash protection
Length (English) Inseam 30 in.
Length (Metric) Inseam 76.2 cm
View More Specs
Alert:
NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40 cal/cm2 should be avoied because of blast hazards caused by arc flash.
Products 36
Catalog Number Mfr. No. Arc Flash Rating Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Arc Flash Rating Color Size Price Quantity & Availability  
19-138-131E
View Documents
Honeywell Salisbury™
ACB10030TW2X
100 cal/cm2 Khaki 2X-Large
Each for $1,993.00
Add to cart
19-138-131A
View Documents
Honeywell Salisbury™
ACB10030TWS
100 cal/cm2 Khaki Small
Each for $1,814.00
Add to cart
19-138-131F
View Documents
Honeywell Salisbury™
ACB10030TW3X
100 cal/cm2 Khaki 3X-Large
Each for $1,993.00
Add to cart
19-138-131C
View Documents
Honeywell Salisbury™
ACB10030TWL
100 cal/cm2 Khaki Large
Each for $1,876.00
Add to cart
19-138-131B
View Documents
Honeywell Salisbury™
ACB10030TWM
100 cal/cm2 Khaki Medium
Each for $1,814.00
Add to cart
19-138-131D
View Documents
Honeywell Salisbury™
ACB10030TWXL
100 cal/cm2 Khaki X-Large
Each for $1,814.00
Add to cart
19-138-120C
View Documents
Honeywell Salisbury™
ACB1130BLL
11 cal/cm2 Navy Blue Large
Each for $567.41
Add to cart
19-138-120F
View Documents
Honeywell Salisbury™
ACB1130BL3X
11 cal/cm2 Navy Blue 3X-Large
Each for $633.07
Add to cart
19-138-120A
View Documents
Honeywell Salisbury™
ACB1130BLS
11 cal/cm2 Navy Blue Small
Each for $567.41
Add to cart
19-138-120B
View Documents
Honeywell Salisbury™
ACB1130BLM
11 cal/cm2 Navy Blue Medium
Each for $567.41
Add to cart
19-138-120D
View Documents
Honeywell Salisbury™
ACB1130BLXL
11 cal/cm2 Navy Blue X-Large
Each for $567.41
Add to cart
19-138-120E
View Documents
Honeywell Salisbury™
ACB1130BL2X
11 cal/cm2 Navy Blue 2X-Large
Each for $633.07
Add to cart
19-138-123E
View Documents
Honeywell Salisbury™
ACB2030BL2X
20 cal/cm2 Navy Blue 2X-Large
Each for $718.80
Add to cart
19-138-123F
View Documents
Honeywell Salisbury™
ACB2030BL3X
20 cal/cm2 Navy Blue 3X-Large
Each for $718.80
Add to cart
19-138-123A
View Documents
Honeywell Salisbury™
ACB2030BLS
20 cal/cm2 Navy Blue Small
Each for $662.50
Add to cart
19-138-123B
View Documents
Honeywell Salisbury™
ACB2030BLM
20 cal/cm2 Navy Blue Medium
Each for $662.50
Add to cart
19-138-125F
View Documents
Honeywell Salisbury™
ACB3130RB3X
31 cal/cm2 Royal Blue 3X-Large
Each for $657.50
Add to cart
19-138-125E
View Documents
Honeywell Salisbury™
ACB3130RB2X
31 cal/cm2 Royal Blue 2X-Large
Each for $657.50
Add to cart
19-138-123C
View Documents
Honeywell Salisbury™
ACB2030BLL
20 cal/cm2 Navy Blue Large
Each for $662.50
Add to cart
19-138-123D
View Documents
Honeywell Salisbury™
ACB2030BLXL
20 cal/cm2 Navy Blue X-Large
Each for $662.50
Add to cart
19-138-125C
View Documents
Honeywell Salisbury™
ACB3130RBL
31 cal/cm2 Royal Blue Large
Each for $600.00
Add to cart
19-138-125B
View Documents
Honeywell Salisbury™
ACB3130RBM
31 cal/cm2 Royal Blue Medium
Each for $600.00
Add to cart
19-138-125D
View Documents
Honeywell Salisbury™
ACB3130RBXL
31 cal/cm2 Royal Blue X-Large
Each for $600.00
Add to cart
19-138-125A
View Documents
Honeywell Salisbury™
ACB3130RBS
31 cal/cm2 Royal Blue Small
Each for $600.00
Add to cart
19-138-127B
View Documents
Honeywell Salisbury™
ACB4030GYM
40 cal/cm2 Gray Medium
Each for $674.00
Add to cart
19-138-127C
View Documents
Honeywell Salisbury™
ACB4030GYL
40 cal/cm2 Gray Large
Each for $903.03
Add to cart
19-138-127E
View Documents
Honeywell Salisbury™
ACB4030GY2X
40 cal/cm2 Gray 2X-Large
Each for $735.00
Add to cart
19-138-127F
View Documents
Honeywell Salisbury™
ACB4030GY3X
40 cal/cm2 Gray 3X-Large
Each for $735.00
Add to cart
19-138-127A
View Documents
Honeywell Salisbury™
ACB4030GYS
40 cal/cm2 Gray Small
Each for $674.00
Add to cart
19-138-127D
View Documents
Honeywell Salisbury™
ACB4030GYXL
40 cal/cm2 Gray X-Large
Each for $903.03
Add to cart
19-138-129A
View Documents
Honeywell Salisbury™
ACB7530GYS
75 cal/cm2 Gray Small
Each for $1,077.00
Add to cart
19-138-129C
View Documents
Honeywell Salisbury™
ACB7530GYL
75 cal/cm2 Gray Large
Each for $1,077.00
Add to cart
19-138-129D
View Documents
Honeywell Salisbury™
ACB7530GYXL
75 cal/cm2 Gray X-Large
Each for $1,077.00
Add to cart
19-138-129F
View Documents
Honeywell Salisbury™
ACB7530GY3X
75 cal/cm2 Gray 3X-Large
Each for $1,175.00
Add to cart
19-138-129B
View Documents
Honeywell Salisbury™
ACB7530GYM
75 cal/cm2 Gray Medium
Each for $1,077.00
Add to cart
19-138-129E
View Documents
Honeywell Salisbury™
ACB7530GY2X
75 cal/cm2 Gray 2X-Large
Each for $1,175.00
Add to cart
Description
Description
  • 11 cal/cm2 to 75 cal/cm2 overalls made of arc flash resistant Indura™ Ultra Soft™ fabric
  • 100 cal/cm2 overalls made of TuffWeld™ and Q/9™ material layers
  • All overalls sewn with Nomex™ for added flame protection
  • Relaxed cut for greater mobility with bib front for additional protection
  • 30 in. inseam and adjustable gussets on pant cuffs
  • Integrated heavy-duty suspenders and adjustable hook and pile waist straps
Compliance
Meet ASTM F1506 and NFPA 70E standards.
Specifications
Specifications
ASTM F1506, NFPA 70E™
Flash protection
76.2 cm
Adjustable Hook and Pile Waist Straps
30 in.