Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salisbury™ RUB-OUT™ Hand Cleaner

Safe to use with insulating rubber gloves

Manufacturer:  WH Salisbury & Co. 1451

 View more versions of this product

Catalog No. 19-138-423


  Check Availability
Add to cart

Specifications

Description

  • Non-petroleum-based hand cleaner dissolves and removes grease, oil, ink, tar, paint and more
  • Safe to use with natural rubber or ozone-resistant SALCOR™ rubber
  • Cleans with or without water
  • Contains natural skin conditioners and leaves a fresh citrus scent

Description & Specifications

Specifications

Rub-Out™ Hand Cleaner
For use with Rubber gloves, cleans with or without Water, dissolves grease, oil, ink, tar, paint
Fresh citrus
0.5L
12 Pack
Natural skin conditioners
Rubber gloves
16 oz.
Can
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit