Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Universal Switch Sticks

For shock protection

Supplier:  Honeywell Salisbury™ 4213

 View more versions of this product

Catalog No. 19-138-484


Only null left
Add to Cart

Description

Description

  • Hot sticks with standard splined universal head
  • 1 1/4 in. (3.2cm) dia.
  • Supplied with a prong
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal Switch Stick
3.1 cm
Prong
120 cm
1.2 kg
1.25 in.
Shock protection
48 in.
2.6 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.