Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salivary Amylase Alpha Antibody (5G6F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261862
Description
Salivary Amylase Alpha Monoclonal specifically detects Salivary Amylase Alpha in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Salivary Amylase Alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
1,4-alpha-D-glucan glucanohydrolase 1, alpha-amylase 1, AMY1, AMY1B, AMY1C, amylase, alpha 1A (salivary), amylase, alpha 1A; salivary, amylase, salivary, alpha-1A, EC 3.2.1.1, glycogenase, Salivary alpha-amylase, salivary amylase alpha 1A | |
Mouse | |
57.8 kDa | |
0.1 mL | |
276 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
5G6F7 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
AMY1A | |
Purified recombinant fragment of human AMY1A (AA: 172-284) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction