Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella Antibody (100/358) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100645630.025MG
Description
Salmonella Monoclonal specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Salmonella | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Mouse | |
Protein A purified | |
RUO | |
Antibody recognizes an antigen common to the various Salmonella strains. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Immunocytochemistry, Immunofluorescence, ELISA | |
100/358 | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Native S. enteritidis (NCTC 5188 9.12:g,m). | |
0.025 mg | |
Primary | |
Bacteria | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction