Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella typhi Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™
Mouse Monoclonal Antibody
$922.00 - $9216.00
Specifications
Antigen | Salmonella typhi |
---|---|
Clone | Clone 1 |
Applications | ELISA, Immunodiffusion, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABX87511MI
|
MilliporeSigma
MABX87511KC |
1 mg |
Each for $922.00
|
|
|||||
MABX85110MI
|
MilliporeSigma
MABX875110KC |
10 mg |
Each for $9,216.00
|
|
|||||
Description
Salmonella typhi Monoclonal antibody specifically detects Salmonella typhi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Salmonella typhi | |
ELISA, Immunodiffusion, Western Blot | |
Unconjugated | |
Infectious Diseases | |
Anti-S. typhi | |
IgG1 | |
Affinity Purified | |
Specific for S. Typhi and S. Paratyphi |
Clone 1 | |
Monoclonal | |
Mouse | |
All species | |
Negative S. typhi Antigen | |
Primary | |
2 to 8°C. For long-term storage, freeze at -20°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title