Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella typhi Mouse anti-All, Unconjugated, Clone: Clone 2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABX87521KC
Description
Salmonella typhi Monoclonal antibody specifically detects Salmonella typhi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Salmonella typhi | |
Monoclonal | |
Unconjugated | |
Mouse | |
Affinity Purified | |
Infectious Diseases | |
Specific for S. Typhi and S. Paratyphi | |
2 to 8°C. For long-term storage, freeze at -20°C. |
ELISA, Immunodiffusion, Western Blot | |
Clone 2 | |
Anti-S. typhi | |
Negative S. typhi Antigen | |
1 mg | |
Primary | |
All species | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction