Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella typhi Mouse anti-All, Unconjugated, Clone: Clone 3, MilliporeSigma™

Mouse Monoclonal Antibody

$784.50 - $7850.00

Specifications

Antigen Salmonella typhi
Applications ELISA, Immunodiffusion, Western Blot
Conjugate Unconjugated
Format Affinity Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABX85310MI
View Documents
MilliporeSigma
MABX875310KC
10 mg
Each for $7,850.00
Add to cart
 
MABX87531MI
View Documents
MilliporeSigma
MABX87531KC
1 mg
Each for $784.50
Add to cart
 
Description

Description

Salmonella typhi Monoclonal antibody specifically detects Salmonella typhi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.
Specifications

Specifications

Salmonella typhi
Unconjugated
Mouse
Anti-S. typhi
Monoclonal
Affinity Chromatography Purified
ELISA, Immunodiffusion, Western Blot
Affinity Purified
All
Negative S. typhi Antigen
Primary
2 to 8°C. For long-term storage, freeze at -20°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit