Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Salt Mixture No. 4164
Contains different variety of salts.
Supplier: MP Biomedicals 0290284902
Description
Salt mixture containing calcium lactate and potassium phosphate.Specifications
Not Autoclavable | |
Yes | |
Non-sterile | |
1 ft. |
Animal Diet | |
2 kg | |
Room Temperature | |
0.3048 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?
Provide Content Correction