Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salt, Non-Iodized, Gram, AquaPhoenix

Click to view available options
Quantity:
12 oz.
7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]
7647-14-5
ClNa
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
5234
CHEBI:26710
sodium;chloride
[Na+].[Cl-]
Physical Form Solid
Grade ACS
Recommended Applications Education
Specific Gravity 2.165
Vapor Pressure 1mmHg at 865°C
Boiling Point 1461°C
Color White
Melting Point 801°C
Quantity 12 oz.
Chemical Name or Material Sodium Chloride, ACS