Learn More
Sam50 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 208241AP
Catalog No. 50-173-3108
Add to cart
Description
Description
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al., 2005 ).
Specifications
Specifications
Sam50
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AXV4, Q8BGH2, Q9Y512
CGI 51, OMP85, Sam50, SAMM50, TOB55, TRG 3, TRG3, YNL026W
Rabbit
IgG
150 μL
-20°C
Primary
25813, 300111, 68653
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SAMM50
Liquid
Samm50
Sam50 Fusion Protein Ag14799
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat