Alfa Aesar™ Samarium Cobalt powder, -30 mesh

Magnet Alloy Additive

Manufacturer: Alfa Aesar™ 1260830

Quantity 250g

 View more versions of this product

Catalog No. AA1260830

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091682
    UN Number UN3089
    Chemical Name or Material Samarium Cobalt powder

    Specifications

    Quantity 250g
    Health Hazard 3 GHS P Statement
    P210-P284a-P261-P280-P342+P311-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    UN Number UN3089
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Sensitivity Air & Moisture Sensitive
    TSCA TSCA
    MDL Number MFCD02091682
    Health Hazard 2 GHS H Statement
    H334-H228-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Flammable solid.
    May cause an allergic skin reaction.
    Mesh Size −30 Mesh
    Avg. Mol. Wt. or Mol. Wt. Range Approx. ratio: Sm:Cu:Fe:Zr:Co; 25.5:5:14:3:52.5 wt%
    Chemical Name or Material Samarium Cobalt powder

    GHS H Statement
    H334-H228-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P284a-P261-P280-P342+P311-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit