Learn More
Alfa Aesar™ Samarium Cobalt powder, -30 mesh
Magnet Alloy Additive
Manufacturer:  Alfa Aesar™ 1260818
Quantity 50g
 View more versions of this product
Catalog No. AA1260818
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD02091682
UN3089
Samarium Cobalt powder
Specifications
Approx. ratio: Sm:Cu:Fe:Zr:Co; 25.5:5:14:3:52.5 wt%
Air & Moisture Sensitive
Powder
UN3089
−30 Mesh
50g
MFCD02091682
Samarium Cobalt powder
Safety and Handling
SDS & Safety Documents
GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.
GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
TSCA : TSCA