Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium foil, 0.127mm (0.005in) thick, 99.9% (REO), Thermo Scientific™

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-19-9
Sm
150.36
MFCD00011233
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
7440-19-9
Sm
150.36
MFCD00011233
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
  • Packaged under argon
Quantity 50 x 50 mm
Weight ≈0.6g/25x25mm
Odor Odorless
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Samarium foil, 0.127mm (0.005 in.) thick
Assay (REO)

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Hazard Class: 4.3; Packaging Group: III

EINECSNumber : 231-128-7

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

RUO – Research Use Only