Alfa Aesar™ Samarium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer: Alfa Aesar™ 12420FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA12420FF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-19-9
    MDL Number MFCD00011233
    Chemical Name or Material Samarium foil, 0.25mm (0.01 in.) thick

    Specifications

    Quantity 25 x 25mm
    Packaging Mylar under Argon
    Odor Odorless
    TSCA TSCA
    Storage Note 1
    • Packaged under argon
    Physical Form Foil
    MDL Number MFCD00011233
    CAS 7440-19-9
    Weight ≈1.2g/25x25mm
    Chemical Name or Material Samarium foil, 0.25mm (0.01 in.) thick
    Assay 99.9%, (REO)

    • Packaged under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit