Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Samarium foil, 0.62mm (0.024 in.) thick, 99.9% (REO)

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-19-9
Sm
150.36
MFCD00011233
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
7440-19-9
Sm
150.36
MFCD00011233
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario,unii-42od65l39f,samarium, elemental,atom,powder,ingot,foil, 3n,chips, 3n,ingot, 3n,acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
  • Packaged under argon
Weight ≈2.9g/25x25mm
Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Samarium foil, 0.62mm (0.024 in.) thick
Assay (REO)

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Hazard Class: 4.3; Packaging Group: III

EINECSNumber : 231-128-7

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures