Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Samarium(II) iodide, 0.07 - 0.12M in THF, stab.

Manufacturer:  Alfa Aesar™ 22360AP

Quantity 500mL

 View more versions of this product

Catalog No. AA22360AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

32248-43-4
MFCD00058873
Samarium(II) iodide, Stabilized
404.21
UN3271

Specifications

32248-43-4
Liquid
MFCD00058873
0.07 to 0.12M in THF
Ether-like
Samarium(II) iodide, Stabilized
0.922g/mL
404.21
UN3271
500mL
Air & Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H260-H314-H318
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ETHERS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit