Alfa Aesar™ Samarium(III) acetate hydrate, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1460414

Quantity 25g

 View more versions of this product

Catalog No. AA1460414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17829-86-6
    Formula Weight 327.48 (Anhydrous)
    MDL Number MFCD00150124
    Chemical Name or Material Samarium(III) acetate hydrate

    Specifications

    Formula Weight 327.48 (Anhydrous)
    Odor Odorless
    Physical Form Crystalline Aggregates
    MDL Number MFCD00150124
    CAS 17829-86-6
    Boiling Point 300°C
    Density 1.94g/mL
    TSCA TSCA
    Sensitivity Hygroscopic
    Quantity 25g
    Solubility Information Soluble in water
    Chemical Name or Material Samarium(III) acetate hydrate
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit