Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Samarium(III) chloride hydrate, REacton™, 99.99% (REO)

Click to view available options
Quantity:
5g
25g
100g
10361-82-7
MFCD00011235
10361-82-7
MFCD00011235
Merck Index 14,8347
Solubility Information Soluble in water.
Formula Weight 256.71 (Anhydrous)
Physical Form Wet Crystalline Aggregates
Percent Purity 99.99%
Grade Reacton™
Odor Acidic
Sensitivity Hygroscopic
Melting Point 110°C (in -5H2O)
Quantity 5g
Chemical Name or Material Samarium(III) chloride hydrate
Assay (REO)
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 233-797-0

TSCA : Yes

Recommended Storage : Ambient temperatures