Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Samarium(III) isopropoxide, 99% (REO), 5% w/v in toluene/isopropanol

Precursor for CVD and Sol-Gel

Manufacturer:  Alfa Aesar™ 41311AC

Quantity 25mL

 View more versions of this product

Catalog No. AA41311AC


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

3504-40-3
327.5
UN3274
Sm[OCH(CH3)2]3
MFCD00058820
Samarium(III) isopropoxide

Specifications

3504-40-3
Sm[OCH(CH3)2]3
MFCD00058820
5% w/v in toluene/isopropanol
Alcohol-like
Samarium(III) isopropoxide
Liquid
327.5
UN3274
25mL
Moisture Sensitive
99%, (REO)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit