Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-4408156
Click to view available options
Quantity:
25 g
100 g
13759-83-6
H12N3O15Sm
444.46
MFCD00149857
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
203081
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
13759-83-6
H12N3O15Sm
444.46
MFCD00149857
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
203081
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

It is used in NMR spectroscopy for spectra resolution and is used for doping purpose. It is also used as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 78°C to 79°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water and acetonitrile.
IUPAC Name samarium(3+) hexahydrate trinitrate
Formula Weight 444.45 (336.36 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow crystals
Density 2.375
Chemical Name or Material Samarium(III) nitrate hexahydrate
Show More Show Less
Hazard Category H272-H315-H320-H335
Precautionary Statement P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 233-798-6
RTECSNumber VP3150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit