Samarium(III) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3627809

Quantity 10g

 View more versions of this product

Catalog No. AA3627809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-58-1
    Physical Form 3 to 12mm Sintered Lump
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula Sm2O3
    Formula Weight 348.7
    MDL Number MFCD00011237
    Chemical Name or Material Samarium(III) oxide

    Specifications

    Physical Form 3 to 12mm Sintered Lump
    Formula Weight 348.7
    Quantity 10g
    Color Off-white
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula Sm2O3
    Appearance Off-white 3-12mm Sintered Lump
    MDL Number MFCD00011237
    CAS 12060-58-1
    Chemical Name or Material Samarium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit