Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) oxide, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1122914

Quantity 25g

 View more versions of this product

Catalog No. AA1122914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12060-58-1
99.9% (REO)
348.7
Samarium(III) oxide
Powder
Sm2O3
MFCD00011237

Specifications

12060-58-1
99.9% (REO)
348.7
MFCD00011237
Off-white
Samarium(III) oxide
Powder
Sm2O3
Off-white powder
8.347g/cm3 at 20°C
25g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 235-043-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit