Alfa Aesar™ Samarium pieces, 12mm (0.47 in.) & down, 99.9% (REO)

Manufacturer: Alfa Aesar™ 0060903

Quantity 1g

 View more versions of this product

Catalog No. AA0060903

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-19-9
    MDL Number MFCD00011233
    Chemical Name or Material Samarium pieces

    Specifications

    Quantity 1g
    Packaging Poly bottle
    MDL Number MFCD00011233
    Color Gray
    Odor Odorless
    TSCA TSCA
    Physical Form Pieces
    CAS 7440-19-9
    Storage Note 1
    • Packaged in air stable poly bottle
    Particle Size 12mm (0.47 in.) & down
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Samarium pieces
    Assay 99.9%, (REO)

    • Packaged in air stable poly bottle

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit