Alfa Aesar™ Samarium powder, <250 micron, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1242214

Quantity 25g

 View more versions of this product

Catalog No. AA1242214

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-19-9
    MDL Number MFCD00011233
    UN Number UN3089
    Chemical Name or Material Samarium powder
    Grade Reacton™

    Specifications

    Quantity 25g
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011233
    UN Number UN3089
    Health Hazard 2 GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Particle Size <250 Micron
    Physical Form Powder
    CAS 7440-19-9
    Odor Odorless
    TSCA TSCA
    Health Hazard 1 Danger
    Color Gray to silver
    Chemical Name or Material Samarium powder
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H228-H260
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.

    GHS P Statement
    P210-P231+P232-P280-P370+P378b-P402+P404-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: Special powder for metal fires.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit