Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)

Manufacturer:  Alfa Aesar™ 40298HF

Quantity 10mm

 View more versions of this product

Catalog No. AA40298HF


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-19-9
MFCD00011233
Samarium rod, 12.7mm (0.5 in.) dia.

Specifications

≈0.96g/mm
7440-19-9
Mylar under Argon
MFCD00011233
99.9%, (metals basis excluding Ta)
10mm
Odorless
Cast Rod, dia. is approximate
Samarium rod, 12.7mm (0.5 in.) dia.

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit