Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Samarium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
Description
- Packaged under argon
Specifications
Weight | ≈0.96g/mm |
Quantity | 10 mm |
Odor | Odorless |
Packaging | Mylar under Argon |
Physical Form | ≈0.96g/mm,Cast Rod,diameter is approximate |
Percent Purity | 99.9% |
Chemical Name or Material | Samarium rod, 12.7mm (0.5 in.) dia. |
Assay | (metals basis excluding Ta) |
Safety and Handling
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260
DOTInformation : Hazard Class: 4.3; Packaging Group: III
EINECSNumber : 231-128-7
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures