Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room Available on GSA/VA Contract for Federal Government customers only.
Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room.
Manufacturer: Thermo Scientific™ 07 0005
 View more versions of this product
Catalog No. 13-711-76
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description Containers with caps
    Closure Color White
    Size (Closure) 53mm
    No. per Pack 100
    No. per Case 100
    Capacity (Metric) 120mL
    Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room. These shatter-resistant containers are manufactured from virgin polypropylene and have a proprietary, all plastic, leak-resistant seal that prevents bio-hazardous spills. Bio-Tite containers are tested at varying pressures, making them ideal for use in various transport systems.
    Features:
    • Dual-threaded caps for leak resistant seal
    • Sterile, individually wrapped, peel pouches for easy opening onto a sterile field
    • Available in sterile, economical baggy wrap
    Ordering Information:
    • Case of 100
  • Description & Specifications
Related Products
Product Content Feedback