Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room.

Manufacturer:  Thermo Scientific™ 07 0007

 View more versions of this product

Catalog No. 22-610-134

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room. These shatter-resistant containers are manufactured from virgin polypropylene and have a proprietary, all plastic, leak-resistant seal that prevents bio-hazardous spills. Bio-Tite containers are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps for leak resistant seal
  • Sterile, individually wrapped, peel pouches for easy opening onto a sterile field
  • Available in sterile, economical baggy wrap

Ordering Information:

  • Case of 100
Specifications

Specifications

90mL
48 mm
Sterile
90 mL
Threaded Cap
48mm
100
Yes
Cylinder
Specimen Container
Blue
Containers with caps
100
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.