Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room.

$128.20

Specifications

Description Containers with caps
No. per Case 100
No. per Pack 100
View More Specs

Products 2
Catalog Number Mfr. No. Closure Color Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Closure Color Capacity (Metric) Price Quantity & Availability  
13-711-65 Edge
Accessories View Documents
Thermo Scientific™
07 0006
Blue 120mL
Case of 100 for $128.20
Edge
Add to cart
 
22-610-134
View Documents
Thermo Scientific™
07 0007
Blue 90mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for the Operating Room. These shatter-resistant containers are manufactured from virgin polypropylene and have a proprietary, all plastic, leak-resistant seal that prevents bio-hazardous spills. Bio-Tite containers are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps for leak resistant seal
  • Sterile, individually wrapped, peel pouches for easy opening onto a sterile field
  • Available in sterile, economical baggy wrap

Ordering Information:

  • Case of 100
Warranty: 90 days
Specifications

Specifications

Containers with caps
100
100
SDS

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit