Learn More
Thermo Scientific™ Samco™ Pathology and General Use Specimen Containers
Handle dry or liquid specimens with economical and disposable Thermo Scientific? Samco? Pathology and General Use Specimen Containers. For single-time use.
Manufacturer:  Thermo Scientific™ 4A 0250
 View more versions of this product
Catalog No. 22-610-121
  Check Availability
Add to cart
Specifications
Description
Handle dry or liquid specimens with economical and disposable Thermo Scientific™ Samco™ Pathology and General Use Specimen Containers. For single-time use.
Features:
  • Containers made of virgin polypropylene—resist cracking, splitting
  • Highly resistant to most chemicals including formaldehyde
  • Molded graduations for fill line verification
  • Translucent containers allow for viewing container contents
  • Secure fitting 70mm white ribbed cap with F-217 liner is a general purpose cap design for broad applications
Description & Specifications
Specifications
No
250mL
250mL; Nonsterile; Screw Cap; Bulk
200
Virgin Polypropylene
Specimen container, w/o cap
No
200