Learn More
SAMHD1 Mouse anti-Human, Clone: OTI2D7, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA501954
Catalog No. 50-167-3050
Add to cart
Description
Description
This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.
Specifications
Specifications
SAMHD1
Monoclonal
Unconjugated
SAMHD1
Liquid
SAMHD1
Full length human recombit protein of human SAMHD1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2D7
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y3Z3
CHBL2, DCIP, HDDC1, MOP-5, SBBI88
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25939