Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMHD1 Mouse anti-Human, HRP, Clone: 851, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243689H

Catalog No. NBP243689H

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

SAMHD1
851
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SAMHD1
Recombinant protein encompassing a sequence within the center region of human SAMHD1. The exact sequence is proprietary.
Protein G purified
Stem Cell Markers
Primary
25939
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS with No Preservative
DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit