Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAMM50 Recombinant Rabbit Monoclonal Antibody (JE61-49)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544861
This item is not returnable.
View return policy
Description
SAMM50 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
Specifications
SAMM50 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6AXV4, Q8BGH2, Q9Y512 | |
Samm50 | |
Synthetic peptide within human SAMM50 aa 341-390/469. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
JE61-49 | |
Unconjugated | |
Samm50 | |
1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W | |
Rabbit | |
Protein A | |
RUO | |
25813, 300111, 68653 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction