Learn More
Cole-Parmer Cole-Parmer Sampling Bulb, natural rubber, gray, 2 mL, 10/pk  DFS Item
Manufacturer:  Cole-Parmer 4422202
These sturdy, re-usable and easy-to-use natural rubber bulbs can be used with Pasteur pipettes and small pipettes. Bulbs have a thickened wall to grip the glass firmly and protect it.
Catalog No. NC0917038