Learn More
Supply SolutionsSupplier Diversity Partner Hach Lab Turbidimeter Sample Cells. Material: Glass; Capacity: 30ml.; Material: Glass; Capacity: 30ml DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2084900
Replacement sample cells for use with 18900, 2100N, 2100AN, and ratio laboratory turbidimeters. Caps are included.; Material: Glass; Capacity: 30ml; Material: Glass; Footnote: For use with Hach Ratio 18900, 2100N and 2100AN Laboratory Turbidmeters.; Includes: 6 vials with caps; Sample Volume: 30 mL; Path Length: 25 mm
Catalog No. 50-476-324