Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Sampling Squares
Easily quantify the results of high school field studies.
$40.00
Specifications
Type | Sampling Square |
---|---|
For Use With (Application) | Ideal to quantify the results of high school field studies and can be used on land or in water |
Includes | 4 PVC legs, 2 PVC T connectors, shock cord |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
S68345
![]() |
Science First![]() 6226120 |
1 m |
Each for $40.00
|
N/A |
Description
- Four PVC legs connected with two PVC T-connectors
- Lightweight, plastic device cleanly and quickly marks off either one-quarter square meter (0.25m2) or one square meter (1.0m2)
- When unfolded, it pops into place using a durable shock cord inside the PVC pipe legs
- T-connectors allow the frame to sink for use in water studies
- Collapsible for transport and storage
- Shock cord is contained inside the pipe legs
Specifications
Sampling Square | |
Ideal to quantify the results of high school field studies and can be used on land or in water | |
4 PVC legs, 2 PVC T connectors, shock cord |