Learn More
MilliporeSigma™ Supelco™ Sampling and Switching Valve
Use to connect the LC and GC instruments for fast and dependable introduction of large sample volumes onto a capillary GC column
$1080.00 - $1410.00
Specifications
For Use With (Equipment) LC and GC Instruments
For Use With (Application) Allows for Fast and Dependable Introduction of Large Sample Volumes Onto a Capillary GC Column
Type Sampling/Switching Valve
View More Specs
Products 9
Catalog Number Mfr. No. Size Temperature (Metric) Maximum Price Quantity & Availability  
Catalog Number Mfr. No. Size Temperature (Metric) Maximum Price Quantity & Availability  
11-100-3059
View Documents
MilliporeSigma™ Supelco™
22941
1/16 in. 300°C Each for $1,080.00
Add to cart
11-100-3027
View Documents
MilliporeSigma™ Supelco™
22976
1/16 in. 175°C Each for $1,240.00
Add to cart
11-100-3020
View Documents
MilliporeSigma™ Supelco™
22950
1/16 in. 300°C Each for $1,310.00
Add to cart
11-100-3045
View Documents
MilliporeSigma™ Supelco™
22975
1/8 in. 175°C Each for $1,160.00
Add to cart
11-100-3040
View Documents
MilliporeSigma™ Supelco™
22914
1/8 in. 300°C Each for $1,200.00
Add to cart
11-100-3021
View Documents
MilliporeSigma™ Supelco™
22977
1/8 in. 175°C Each for $1,310.00
Add to cart
11-100-3019
View Documents
MilliporeSigma™ Supelco™
22915
1/8 in. 300°C Each for $1,330.00
Add to cart
11-100-3008
View Documents
MilliporeSigma™ Supelco™
22916
1/8 in. 300°C Each for $1,410.00
Add to cart
11-100-3009
View Documents
MilliporeSigma™ Supelco™
22981
1/8 in. 175°C Each for $1,410.00
Add to cart
Description
Description
  • Supports discrete volumes of the treated sample's stream to periodically inject into the carrier gas stream of the chromatograph
  • Fitted with electric or pneumatic actuators to help with automatic operation
  • Consists of 5 parts: control unit, connecting device, valve body, rotor, and preloaded assembly
  • Isolate a column to prevent a compound emerging from an upstream column, from being irreversibly adsorbed
  • Valve allows employment of different, desired mobile phases
  • Critical when binary pumps are used
Specifications
Specifications
LC and GC Instruments
Allows for Fast and Dependable Introduction of Large Sample Volumes Onto a Capillary GC Column
Sampling/Switching Valve