Learn More
SAMSN1 Mouse anti-Human, Clone: OTI1F6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808889
Catalog No. 50-166-9153
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SAMSN (HACS1) is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains. It encodes a 441 amino acid protein that is differentially expressed in hematopoietic cells and maligcies including myeloid leukemia, lymphoma, and myeloma. SAMSN has restricted expression in human tissues. It has been shown to be up-regulated by B cell activation signals and is a participant in B cell activation and differentiation. SAMSN is an immunoinhibitory adaptor that might be a useful target for immune suppression therapy.
Specifications
Specifications
SAMSN1
Monoclonal
Unconjugated
SAMSN1
Lyophilized
SAMSN1
Human recombit protein fragment corresponding to amino acids 1-250 of human SAMSN1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1F6
PBS with 8% trehalose and no preservative; pH 7.3
Q9NSI8
HACS1, NASH1, SASH2, SH3D6B, SLy2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64092