Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sand, Ottawa, Standard, 20-30 Mesh, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
14808-60-7
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O
14808-60-7
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O

S1010, 14808-60-7

Spectrum™ Sand, Ottawa, Standard, 20-30 Mesh
Grade Reagent
CAS Max % 1
Packaging Fiber Drum
Quantity 45 kg