Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sand, washed, Thermo Scientific Chemicals

Catalog Number p-7052616
Click to view available options
Quantity:
1000 g
5000 g
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O

Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sand, washed is employed in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes. It may be employed as insulating layer on top of the silicon in the fabrication of gold electrode, used for flow injection amperometric detection of organophosphorus (OP) nerve agents.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Boiling Point 2230°C
Melting Point 1710°C
Color Brown-Cream
Physical Form Granules
Quantity 1000 g
Linear Formula SiO2
Solubility Information Insoluble in water.
Formula Weight 60.08
Odor Odorless
Chemical Name or Material Sand, Washed
Hazard Category H350
Hazard Statement GHS H Statement
H350-H373
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 238-878-4
RTECSNumber W7330000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit