Learn More
Eisco™ Sandstone Specimen Products for Science Education
Sandstone Specimen is made up of well-sorted and well-rounded grains of quartz.
$.36 - $28.00
Save up to 75%
Specifications
Specimen Type Sandstone
Type Sedimentary Rock
View More Specs
Products 4
Catalog Number Mfr. No. Specimen Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Specimen Size Quantity Price Quantity & Availability  
S24738 Products for Science Education View Documents Eisco™
ESNG0036
2 to 3 cm 1 Ea. Each for $0.36
Save 75% Reg : $1.44
Add to cart
S26665 Products for Science Education View Documents Eisco™
ESNG0036PK10
2 to 3 cm 10/Pk. Each for $2.60
Add to cart
S24739 Products for Science Education View Documents Eisco™
ESNG0036PK12
2 to 3 cm 12/Pk Pack of 12 for $28.00
Add to cart
S26682 Products for Science Education View Documents Eisco™
ESNG0036HAND
7 to 8 cm 1 Each Each for $13.00
Add to cart
Description
Description
  • For use in studying depositional environments
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Rounded, well-sorted sand grains are excellent identifiers for students during sedimentary rock identification
  • This indicates that clasts in rocks have traveled long distance from their source and were heavily eroded along way
Specifications
Specifications
Sandstone
Sedimentary Rock