Learn More
V&P Scientific Inc STIR ELEMENT, SANDWICH STYLE V SHAPE, NdFeB Magnet, 42MGO, PTFE Plate, Used in Microplates, Tubes/Vials, 10/pack
Supplier:  V&P Scientific Inc VP 772FN515V150
Encompass
STIR ELEMENT, SANDWICH STYLE V SHAPE, NdFeB Magnet, 42MGO, PTFE Plate, 15.0mm Length, 5.0mm Width, 1.6mm Height, 150°C Max Operating Temp for Magnets
Catalog No. NC1653036