Learn More
Saint-Gobain Sani-Tech™ SIL-250 Extended Life Silicone Tubing
Sani-Tech™ SIL-250 is a high-performance, extended-life, platinum-cured silicone tubing formulation specifically designed for demanding peristaltic pump applications. Its superior flex life characteristics reduce potential production downtime due to pump tubing failure.
Manufacturer:  Saint-Gobain F90016
 View more versions of this product
Catalog No. 13-417-206
Add to cart
Description
Description
Sani-Tech™ SIL-250 has an extremely smooth inner surface that helps minimize the risk of particle entrapment during sensitive fluid transfer.
  • Lasts up to five times longer than other platinum silicone tubing
  • Minimal extractables help maintain fluid integrity
  • Ultra-smooth inner bore reduces potential particle entrapment
  • Documented biocompatibility for sensitive applications
  • Excellent fluid flow characteristics
  • Taste-free and odor-free
  • Meets USP Class VI and FDA criteria
Specifications
Specifications
USP Class VI, FDA 21 CFR Part 177.2600
Autoclavable
7/16 in.
15.24 m
400%
Peristaltic pumps
400°F
SIL-250/187-4
Translucent
3/16 in.
50 ft.
0.125 in.
Bend Radius 3/8 in.
55 (Shore A)
204°C