Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP102 Mouse anti-Human, Mouse, Rat, Clone: S19-2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527707

Catalog No. PIMA527707


Add to cart

Description

Description

Synapse-Associated Protein 102 (SAP102) is one of a family of plasma membrane-associated proteins found in synaptic junctions. Like other members of the family, SAP102 has three ~90 amino acid repeats called PDZ domains followed by an SH3 domain and a yeast guanylate kinase homology (GuK) domain. It is hypothesized that PDZ-domain interactions play a role in receptor and channel clustering which contributes to neuronal plasticity. SAP102 is believed to participate in the clustering of certain proteins, including NMDA receptors, shaker-type potassium channels at the synaptic membrane in CNS neurons. NMDA receptors and Shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif (beta 1 adrenergic receptor, some serotonin receptors, some sodium channel subunits, and additional potassium channel subunits) may interact with SAP102 by binding to its PDZ domains. Neuronal nitric oxide synthase (nNOS), which lacks the T/SXV motif but which has its own PDZ domain, has been shown to associate with SAP102 in vitro through a pseudo-homotypic PDZ-PDZ interaction.
Specifications

Specifications

SAP102
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P70175, Q62936, Q92796
DLG3, MRX90, NEDLG, XLMR, Disks large homolog 3, PSD-95/SAP90-related protein 1, Synapse-associated protein 102, SAP-102
Mouse
IgG1
100 μg
-20°C
Primary
1741, 53310, 58948
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
S19-2
Unconjugated
DLG3
Liquid
DLG3
Fusion protein amino acids 1-120 of rat SAP102
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit