Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAP102 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S19-2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP121672IR
Description
SAP102 Monoclonal antibody specifically detects SAP102 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
SAP102 | |
S19-2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q62936 | |
discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR | |
Mouse | |
IgG1 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
Detects approx 105kDa. No cross-reactivity against PSD95, SAP97 or Chapsyn-110. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified | |
DLG3 | |
Fusion protein amino acids 1-120 of rat SAP102 | |
Protein G purified | |
Cell Cycle and Replication, Neuroscience | |
Primary | |
1741 | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title