Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
SAP102 Mouse anti-Human, Mouse, Rat, HRP, Clone: S19-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121672H
Catalog No. NBP121672H
Description
Description
SAP102 Monoclonal antibody specifically detects SAP102 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
SAP102
S19-2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Q62936
discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 105kDa. No cross-reactivity against PSD95, SAP97 or Chapsyn-110.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
Purified
DLG3
Fusion protein amino acids 1-120 of rat SAP102
Protein G purified
Cell Cycle and Replication, Neuroscience
Primary
1741
Human, Mouse, Rat